Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004947134.3 | XP_004947191.1 | 3 - 4 | 663 | 106 | |
XM_417434.6 | XP_417434.2 | 3 - 4 | 682 | 106 | ||
GenBank mRNA | AY251406.1 | - | 2 - 3 | 415 | 106 | |
BX931790.1 | - | 2 - 3 | 623 | 106 | ||
BX931818.1 | - | 1 - 2 | 332 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons |