Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_419794.5 | XP_419794.2 | 7 - 8 | 791 | 66 | |
GenBank mRNA | BX929326.1 | - | 6 - 7 | 547 | 66 | |
CR390919.1 | - | 6 - 7 | 691 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |