Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNIH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015283910.2 | XP_015139396.1 | 3 - 4 | 270 | 96 | |
XM_025149053.1 | XP_025004821.1 | 3 - 4 | 292 | 96 | ||
XM_419390.6 | XP_419390.1 | 3 - 4 | 288 | 96 | ||
GenBank mRNA | BX929407.1 | - | 3 - 4 | 301 | 96 | |
BX950810.1 | - | 1 - 2 | 402 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Gallus gallus Probe spans exons |