Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DZANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199396.1 | NP_001186325.1 | 2 - 3 | 240 | 77 | |
XM_015283472.2 | XP_015138958.1 | 2 - 3 | 344 | 77 | ||
XM_015283473.2 | XP_015138959.1 | 2 - 3 | 344 | 77 | ||
XM_025148594.1 | XP_025004362.1 | 2 - 3 | 391 | 77 | ||
GenBank mRNA | BX929649.1 | - | 1 - 2 | 41 | 77 | |
BX933366.2 | - | 2 - 3 | 241 | 77 | ||
CR407362.1 | - | 2 - 3 | 394 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |