Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC51B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015292256.2 | XP_015147742.1 | 3 - 4 | 943 | 57 | |
XM_025153900.1 | XP_025009668.1 | 3 - 4 | 626 | 57 | ||
XM_025153901.1 | XP_025009669.1 | 3 - 4 | 2573 | 57 | ||
GenBank mRNA | BX929774.1 | - | 4 - 5 | 754 | 57 | |
BX930136.2 | - | 2 - 3 | 734 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons |