Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198655.3 | NP_001185584.1 | 3 - 4 | 442 | 79 | |
GenBank mRNA | BX929792.1 | - | 3 - 4 | 335 | 79 | |
BX932521.1 | - | 3 - 4 | 443 | 79 | ||
BX934036.2 | - | 2 - 3 | 201 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |