Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MICU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_417140.6 | XP_417140.6 | 4 - 5 | 469 | 96 | |
GenBank mRNA | BX929803.1 | - | 4 - 5 | 465 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |