Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MICU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_417140.6 | XP_417140.6 | 7 - 8 | 672 | 141 | |
GenBank mRNA | BX929803.1 | - | 7 - 8 | 668 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |