Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277910.3 | NP_001264839.1 | 1 - 2 | 626 | 77 | |
XM_025142529.1 | XP_024998297.1 | 4 - 5 | 681 | 77 | ||
XM_025142530.1 | XP_024998298.1 | 6 - 7 | 839 | 77 | ||
GenBank mRNA | BX929880.1 | - | 1 - 2 | 202 | 77 | |
BX934498.1 | - | 1 - 2 | 500 | 77 | ||
BX950638.1 | - | 1 - 2 | 633 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |