Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252162.1 | NP_001239091.1 | 2 - 3 | 252 | 235 | |
GenBank mRNA | BX929945.1 | - | 2 - 3 | 181 | 100 | |
BX933305.1 | - | 1 - 2 | 171 | 235 | ||
CR353075.1 | - | 1 - 2 | 121 | 235 |
Set Membership: |
Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |