Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285926.1 | NP_001272855.1 | 3 - 4 | 253 | 82 | |
XM_004938247.3 | XP_004938304.1 | 3 - 4 | 337 | 82 | ||
XM_004938248.3 | XP_004938305.1 | 3 - 4 | 253 | 82 | ||
GenBank mRNA | BX930121.1 | - | 3 - 4 | 250 | 82 | |
BX930142.2 | - | 4 - 5 | 400 | 82 | ||
BX930156.1 | - | 4 - 5 | 400 | 82 | ||
BX930576.2 | - | 3 - 4 | 250 | 82 | ||
BX930906.1 | - | 3 - 4 | 263 | 82 | ||
BX931243.2 | - | 3 - 4 | 250 | 82 | ||
BX933264.1 | - | 3 - 4 | 233 | 82 | ||
BX935375.1 | - | 3 - 4 | 253 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |