Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025151664.1 | XP_025007432.1 | 3 - 4 | 441 | 96 | |
XM_421730.6 | XP_421730.1 | 3 - 4 | 335 | 96 | ||
GenBank mRNA | BX929388.1 | - | 2 - 3 | 287 | 96 | |
BX930620.1 | - | 3 - 4 | 329 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |