Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR7A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_417628.6 | XP_417628.2 | 4 - 5 | 610 | 75 | |
GenBank mRNA | BX930804.2 | - | 4 - 5 | 483 | 75 | |
BX930844.1 | - | 4 - 5 | 600 | 75 | ||
BX931408.1 | - | 4 - 5 | 579 | 75 | ||
BX934026.2 | - | 4 - 5 | 602 | 75 | ||
BX934046.1 | - | 4 - 5 | 602 | 75 | ||
CR353499.1 | - | 4 - 5 | 597 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |