Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278076.1 | NP_001265005.1 | 6 - 7 | 758 | 88 | |
NM_001278077.1 | NP_001265006.1 | 5 - 6 | 678 | 88 | ||
XM_015296397.2 | XP_015151883.2 | 6 - 7 | 6181 | 88 | ||
GenBank mRNA | BU324326.1 | - | 4 - 5 | 491 | 89 | |
BX932077.1 | - | 6 - 7 | 758 | 88 | ||
BX932300.1 | - | 4 - 5 | 540 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |