Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277561.1 | NP_001264490.1 | 1 - 2 | 179 | 74 | |
GenBank mRNA | BU301369.1 | - | 1 - 2 | 179 | 74 | |
BX929859.2 | - | 1 - 2 | 157 | 74 | ||
BX932342.1 | - | 1 - 2 | 51 | 74 | ||
BX932519.1 | - | 1 - 2 | 179 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |