Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199565.1 | NP_001186494.1 | 3 - 4 | 282 | 66 | |
NM_001199566.1 | NP_001186495.1 | 3 - 4 | 282 | 66 | ||
XM_015286220.2 | XP_015141706.2 | 3 - 4 | 283 | 66 | ||
XM_025150660.1 | XP_025006428.1 | 3 - 4 | 803 | 66 | ||
XM_025150661.1 | XP_025006429.1 | 3 - 4 | 802 | 66 | ||
XM_025150662.1 | XP_025006430.1 | 3 - 4 | 801 | 66 | ||
XM_025150663.1 | XP_025006431.1 | 3 - 4 | 673 | 66 | ||
XM_025150664.1 | XP_025006432.1 | 3 - 4 | 232 | 66 | ||
GenBank mRNA | AM065422.1 | - | 3 - 4 | 282 | 66 | |
BU253847.1 | - | 1 - 2 | 31 | 66 | ||
BX929358.2 | - | 3 - 4 | 275 | 66 | ||
BX932402.1 | - | 3 - 4 | 277 | 66 | ||
BX934566.2 | - | 2 - 3 | 296 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |