Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025142566.1 | XP_024998334.1 | 3 - 4 | 376 | 140 | |
XM_025142567.1 | XP_024998335.1 | 3 - 4 | 376 | 140 | ||
GenBank mRNA | BX932578.1 | - | 3 - 4 | 187 | 140 | |
BX934819.1 | - | 3 - 4 | 189 | 140 |
Gene Symbol: AGTRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015297039.1 | XP_015152525.1 | 3 - 4 | 183 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |