Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199473.1 | NP_001186402.1 | 1 - 2 | 100 | 90 | |
XM_025147090.1 | XP_025002858.1 | 1 - 2 | 146 | 90 | ||
GenBank mRNA | BX930666.2 | - | 1 - 2 | 83 | 90 | |
BX931460.2 | - | 1 - 2 | 83 | 90 | ||
BX932726.1 | - | 1 - 2 | 93 | 90 | ||
CF250315.1 | - | 1 - 2 | 102 | 90 | ||
CR353353.1 | - | 1 - 2 | 89 | 90 | ||
CR406236.1 | - | 1 - 2 | 84 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons |