Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL20RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_419723.5 | XP_419723.1 | 1 - 2 | 174 | 140 | |
GenBank mRNA | BX933318.1 | - | 1 - 2 | 142 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |