Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL20RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX933318.1 | - | 3 - 4 | 475 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |