Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIEZO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015282541.2 | XP_015138027.2 | 9 - 10 | 1348 | 64 | |
XM_015282543.2 | XP_015138029.2 | 9 - 10 | 1348 | 64 | ||
XM_025147818.1 | XP_025003586.1 | 9 - 10 | 1348 | 64 | ||
XM_025147819.1 | XP_025003587.1 | 8 - 9 | 1236 | 64 | ||
XM_025147820.1 | XP_025003588.1 | 9 - 10 | 1348 | 64 | ||
XM_025147821.1 | XP_025003589.1 | 5 - 6 | 711 | 64 | ||
XM_025147822.1 | XP_025003590.1 | 9 - 10 | 1348 | 64 | ||
XM_025147824.1 | XP_025003592.1 | 9 - 10 | 1348 | 64 | ||
GenBank mRNA | BX931090.2 | - | 3 - 4 | 237 | 64 | |
BX933406.1 | - | 1 - 2 | 119 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |