Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199430.1 | NP_001186359.1 | 4 - 5 | 402 | 72 | |
GenBank mRNA | BI395062.1 | - | 4 - 5 | 402 | 72 | |
BX933953.1 | - | 3 - 4 | 225 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |