Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199430.1 | NP_001186359.1 | 8 - 9 | 74 | ||
GenBank mRNA | BX933953.1 | - | 7 - 8 | 554 | 74 | |
CR386813.1 | - | 2 - 3 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |