Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSGEPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004942397.3 | XP_004942454.1 | 6 - 7 | 1456 | 61 | |
XM_004942398.3 | XP_004942455.1 | 6 - 7 | 1456 | 61 | ||
XM_004942399.3 | XP_004942456.1 | 5 - 6 | 1210 | 61 | ||
XM_025152269.1 | XP_025008037.1 | 6 - 7 | 1456 | 61 | ||
GenBank mRNA | BX930694.2 | - | 4 - 5 | 663 | 61 | |
BX930963.2 | - | 4 - 5 | 1361 | 61 | ||
BX934991.1 | - | 6 - 7 | 1240 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |