Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSGEPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004942397.3 | XP_004942454.1 | 7 - 8 | 1525 | 144 | |
XM_004942399.3 | XP_004942456.1 | 6 - 7 | 1279 | 144 | ||
GenBank mRNA | BX930694.2 | - | 5 - 6 | 732 | 144 | |
BX934991.1 | - | 7 - 8 | 1309 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |