Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYKK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277459.1 | NP_001264388.1 | 4 - 5 | 710 | 72 | |
XM_015278879.2 | XP_015134365.2 | 4 - 5 | 1161 | 72 | ||
XM_015278880.2 | XP_015134366.2 | 4 - 5 | 1166 | 72 | ||
XM_015278881.2 | XP_015134367.1 | 4 - 5 | 784 | 72 | ||
XM_015278882.2 | XP_015134368.1 | 4 - 5 | 806 | 72 | ||
XM_015278883.2 | XP_015134369.1 | 4 - 5 | 693 | 72 | ||
XM_025153828.1 | XP_025009596.1 | 4 - 5 | 2527 | 72 | ||
GenBank mRNA | BX933588.2 | - | 4 - 5 | 710 | 72 | |
BX935072.1 | - | 3 - 4 | 803 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |