Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277576.1 | NP_001264505.1 | 5 - 6 | 446 | 150 | |
XM_015283680.2 | XP_015139166.1 | 6 - 7 | 505 | 150 | ||
GenBank mRNA | BU262047.1 | - | 5 - 6 | 430 | 150 | |
BX930192.1 | - | 5 - 6 | 433 | 150 | ||
BX930686.1 | - | 5 - 6 | 419 | 150 | ||
BX930708.2 | - | 5 - 6 | 423 | 150 | ||
BX933704.1 | - | 6 - 7 | 508 | 150 | ||
BX935171.1 | - | 6 - 7 | 622 | 150 | ||
CD763841.1 | - | 5 - 6 | 435 | 150 | ||
CF255365.1 | - | 5 - 6 | 448 | 150 | ||
CR407465.1 | - | 5 - 6 | 414 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |