Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_417753.6 | XP_417753.1 | 3 - 4 | 325 | 127 | |
GenBank mRNA | BX930871.2 | - | 3 - 4 | 295 | 127 | |
BX935563.1 | - | 3 - 4 | 301 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |