Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_417753.6 | XP_417753.1 | 4 - 5 | 440 | 101 | |
GenBank mRNA | BX930871.2 | - | 4 - 5 | 410 | 101 | |
BX935563.1 | - | 4 - 5 | 416 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |