Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278013.1 | NP_001264942.1 | 1 - 2 | 106 | 82 | |
NM_001278014.1 | NP_001264943.1 | 1 - 2 | 106 | 82 | ||
GenBank mRNA | BM491106.1 | - | 1 - 2 | 77 | 82 | |
BX930908.2 | - | 1 - 2 | 86 | 82 | ||
BX935078.2 | - | 1 - 2 | 72 | 82 | ||
BX936083.1 | - | 1 - 2 | 89 | 82 | ||
CR386502.1 | - | 1 - 2 | 76 | 82 | ||
CR733361.1 | - | 1 - 2 | 89 | 82 | ||
CV891325.1 | - | 1 - 2 | 106 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |