Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277604.1 | NP_001264533.1 | 2 - 3 | 419 | 93 | |
NR_102410.1 | 2 - 3 | 355 | 93 | |||
XM_015283712.2 | XP_015139198.1 | 2 - 3 | 395 | 93 | ||
GenBank mRNA | BU492830.1 | - | 2 - 3 | 301 | 93 | |
BX950488.1 | - | 2 - 3 | 419 | 93 | ||
CN227248.1 | - | 1 - 2 | 48 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |