Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST6GALNAC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277494.1 | NP_001264423.1 | 2 - 3 | 345 | 65 | |
XM_025152773.1 | XP_025008541.1 | 2 - 3 | 656 | 65 | ||
XM_025152774.1 | XP_025008542.1 | 2 - 3 | 910 | 65 | ||
XM_025152775.1 | XP_025008543.1 | 2 - 3 | 649 | 65 | ||
XM_025152776.1 | XP_025008544.1 | 2 - 3 | 656 | 65 | ||
GenBank mRNA | AJ646877.1 | - | 2 - 3 | 247 | 65 | |
CD526958.1 | - | 2 - 3 | 365 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |