Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_421970.6 | XP_421970.3 | 4 - 5 | 505 | 77 | |
GenBank mRNA | BX929596.2 | - | 4 - 5 | 471 | 77 | |
BX929642.2 | - | 4 - 5 | 471 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |