Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELMOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277915.1 | NP_001264844.1 | 7 - 8 | 744 | 144 | |
GenBank mRNA | BX929856.2 | - | 7 - 8 | 714 | 144 | |
BX932331.2 | - | 7 - 8 | 744 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |