Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199421.1 | NP_001186350.1 | 3 - 4 | 1019 | 68 | |
GenBank mRNA | BX930866.2 | - | 2 - 3 | 227 | 68 | |
BX935703.1 | - | 3 - 4 | 913 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |