Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015294578.2 | XP_015150064.1 | 4 - 5 | 580 | 127 | |
XM_025155257.1 | XP_025011025.1 | 4 - 5 | 553 | 127 | ||
XM_414935.6 | XP_414935.1 | 4 - 5 | 526 | 127 | ||
GenBank mRNA | BX930973.2 | - | 4 - 5 | 517 | 127 | |
BX933414.2 | - | 4 - 5 | 539 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |