Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277994.1 | NP_001264923.1 | 5 - 6 | 620 | 123 | |
GenBank mRNA | BX932102.2 | - | 5 - 6 | 620 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |