Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277562.1 | NP_001264491.1 | 3 - 4 | 565 | 76 | |
XM_015276734.2 | XP_015132220.1 | 3 - 4 | 599 | 76 | ||
GenBank mRNA | BU267331.1 | - | 3 - 4 | 487 | 76 | |
BU356089.1 | - | 3 - 4 | 562 | 76 | ||
BU476835.1 | - | 3 - 4 | 479 | 76 | ||
BX273937.4 | - | 3 - 4 | 470 | 76 | ||
BX931650.2 | - | 3 - 4 | 565 | 76 | ||
BX932166.2 | - | 3 - 4 | 553 | 76 | ||
BX932359.2 | - | 3 - 4 | 562 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |