Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_422526.6 | XP_422526.4 | 4 - 5 | 757 | 65 | |
GenBank mRNA | BX930997.2 | - | 5 - 6 | 698 | 65 | |
BX932222.2 | - | 4 - 5 | 492 | 65 | ||
BX933246.1 | - | 3 - 4 | 373 | 65 | ||
CR406857.1 | - | 4 - 5 | 470 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |