Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025151675.1 | XP_025007443.1 | 5 - 6 | 913 | 75 | |
XM_421748.4 | XP_421748.3 | 4 - 5 | 653 | 75 | ||
GenBank mRNA | BX933054.2 | - | 3 - 4 | 808 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |