Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004945487.3 | XP_004945544.1 | 1 - 2 | 318 | 102 | |
GenBank mRNA | BX933718.2 | - | 1 - 2 | 280 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |