Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277141.1 | NP_001264070.1 | 7 - 8 | 736 | 143 | |
XM_015293066.2 | XP_015148552.1 | 8 - 9 | 1014 | 143 | ||
XM_025154462.1 | XP_025010230.1 | 8 - 9 | 832 | 143 | ||
XM_025154463.1 | XP_025010231.1 | 7 - 8 | 568 | 143 | ||
GenBank mRNA | BX933925.2 | - | 3 - 4 | 167 | 143 | |
BX950678.1 | - | 6 - 7 | 598 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |