Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAD2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277749.1 | NP_001264678.1 | 1 - 2 | 111 | 62 | |
GenBank mRNA | BX934016.2 | - | 1 - 2 | 111 | 62 | |
BX935088.1 | - | 1 - 2 | 101 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |