Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004943720.3 | XP_004943777.1 | 2 - 3 | 469 | 70 | |
XM_004943722.3 | XP_004943779.1 | 2 - 3 | 158 | 70 | ||
XM_004943723.3 | XP_004943780.1 | 2 - 3 | 125 | 70 | ||
XM_413749.6 | XP_413749.1 | 2 - 3 | 489 | 70 | ||
GenBank mRNA | BX935149.2 | - | 2 - 3 | 460 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |