Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXNAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199638.1 | NP_001186567.1 | 9 - 10 | 830 | 135 | |
XM_015281479.2 | XP_015136965.1 | 8 - 9 | 988 | 135 | ||
XM_015281480.2 | XP_015136966.1 | 8 - 9 | 1054 | 135 | ||
XM_015281481.2 | XP_015136967.1 | 8 - 9 | 956 | 135 | ||
XM_015281482.2 | XP_015136968.1 | 7 - 8 | 859 | 135 | ||
GenBank mRNA | BU408430.1 | - | 4 - 5 | 606 | 135 | |
BX932544.2 | - | 7 - 8 | 1157 | 135 | ||
BX935163.2 | - | 7 - 8 | 891 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |