Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277728.1 | NP_001264657.1 | 3 - 4 | 349 | 86 | |
GenBank mRNA | BU105412.1 | - | 2 - 3 | 298 | 86 | |
BU222325.1 | - | 3 - 4 | 351 | 86 | ||
BX935079.1 | - | 2 - 3 | 471 | 86 | ||
BX935284.2 | - | 2 - 3 | 303 | 86 | ||
CR386367.1 | - | 2 - 3 | 394 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |