Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_418224.6 | XP_418224.2 | 7 - 8 | 686 | 59 | |
GenBank mRNA | BX929663.1 | - | 7 - 8 | 670 | 59 | |
BX930905.2 | - | 7 - 8 | 669 | 59 | ||
BX935423.2 | - | 7 - 8 | 673 | 59 | ||
BX935442.2 | - | 7 - 8 | 661 | 59 | ||
CR385484.1 | - | 7 - 8 | 649 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |