Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004941776.3 | XP_004941833.1 | 3 - 4 | 483 | 129 | |
XM_004941778.3 | XP_004941835.1 | 3 - 4 | 484 | 129 | ||
XM_015287662.2 | XP_015143148.1 | 3 - 4 | 484 | 129 | ||
GenBank mRNA | BX935451.2 | - | 3 - 4 | 479 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |