Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277613.1 | NP_001264542.1 | 1 - 2 | 165 | 90 | |
GenBank mRNA | BU330715.1 | - | 1 - 2 | 165 | 90 | |
BX933248.1 | - | 1 - 2 | 149 | 90 | ||
BX935707.2 | - | 1 - 2 | 108 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |