Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS7C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277995.1 | NP_001264924.1 | 2 - 3 | 253 | 69 | |
GenBank mRNA | BX929314.1 | - | 2 - 3 | 254 | 69 | |
BX935860.2 | - | 2 - 3 | 269 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |